APO455834418 Apollo Scientific Ltd

2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]aniline

  • MDL number MFCD22628762
  • Cas Number : 156425-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147
Synonym 2-Amino-3-bromo-5-[(trifluoromethyl)sulphonyl]nitrobenzene, 4-Amino-3-bromo-5-nitrophenyl trifluoromethyl sulphone
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51289

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51289
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart