APO455834420 Apollo Scientific Ltd

2-Bromo-5-(trifluoromethoxy)benzyl bromide

  • MDL number MFCD07368721
  • Cas Number : 886763-13-9
  • Molecular Weight : 333.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-(bromomethyl)-4-(trifluoromethoxy)benzene
Inchi Code 1S/C8H5Br2F3O/c9-4-5-3-6(1-2-7(5)10)14-8(11,12)13/h1-3H,4H2
Inchi Key TVBXKZSWLSZTRG-UHFFFAOYSA-N

Description

Synonym 1-Bromo-2-(bromomethyl)-4-(trifluoromethoxy)benzene, 4-Bromo-3-(bromomethyl)phenyl trifluoromethyl ether
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-trifluoromethoxybenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-trifluoromethoxybenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart