APO455834422 Apollo Scientific Ltd

5-Bromo-2-fluorobenzyl bromide

  • MDL number MFCD09038457
  • Cas Number : 99725-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33
Synonym 4-Bromo-2-(bromomethyl)-1-fluorobenzene, alpha,5-Dibromo-2-fluorotoluene
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 126-136/15mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55693

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55693
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart