APO455834451 Apollo Scientific Ltd

2-(Aminomethyl)-3-fluoroaniline

  • MDL number MFCD00042483
  • Cas Number : 175277-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-6-fluorobenzylamine
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 78-80/1mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=100

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=100
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart