APO455834460 Apollo Scientific Ltd

3-Amino-4-(2-methoxyphenoxy)benzotrifluoride

  • MDL number MFCD00052419
  • Cas Number : 175135-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28
Purity 97
Physical Form Solid-Low Melt
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=121

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=121
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart