APO455834462 Apollo Scientific Ltd

2-Amino-3-(methylthiomethyl)benzotrifluoride

  • MDL number MFCD00052842
  • Cas Number : 88301-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Methylthiomethyl)-6-(trifluoromethyl)aniline
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 71-73/6mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=127

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=127
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart