APO455834466 Apollo Scientific Ltd

2-Amino-3-fluorobenzoic acid

  • MDL number MFCD01569395
  • Cas Number : 825-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-187
Synonym 3-Fluoroanthranilic acid, 2-Carboxy-6-fluoroaniline
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-fluorobenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart