APO455834469 Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD04115886
  • Cas Number : 886497-81-0
  • Molecular Weight : 208.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethoxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethoxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart