APO455834489 Apollo Scientific Ltd

3-Bromo-4-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD08458004
  • Cas Number : 85366-66-1
  • Molecular Weight : 269.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-4-(trifluoromethoxy)benzaldehyde
Inchi Code 1S/C8H4BrF3O2/c9-6-3-5(4-13)1-2-7(6)14-8(10,11)12/h1-4H
Inchi Key OCIAOFZMQWTYNX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-trifluoromethoxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-trifluoromethoxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart