APO455834492 Apollo Scientific Ltd

5-Bromo-2-fluoronicotinaldehyde

  • MDL number MFCD09842440
  • Cas Number : 875781-15-0
  • Molecular Weight : 204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-fluoropyridine-3-carboxaldehyde, 5-Bromo-2-fluoro-3-formylpyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-fluoronicotinaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-fluoronicotinaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart