APO455834504 Apollo Scientific Ltd

3'-Fluoro-[1,1'-biphenyl]-4-carboxaldehyde

  • MDL number MFCD04039225
  • Cas Number : 400750-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3'-Fluoro-4-formylbiphenyl, 4-(3-Fluorophenyl)benzaldehyde
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8669

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8669
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart