APO455834506 Apollo Scientific Ltd

3'-Fluoro-[1,1'-biphenyl]-2-carboxylic acid

  • MDL number MFCD03426488
  • Cas Number : 2094-03-3
  • Molecular Weight : 216.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 96-98
Iupac Name 3'-fluoro[1,1'-biphenyl]-2-carboxylic acid
Inchi Code 1S/C13H9FO2/c14-10-5-3-4-9(8-10)11-6-1-2-7-12(11)13(15)16/h1-8H,(H,15,16)
Inchi Key GEZNPQHOTCPFEL-UHFFFAOYSA-N

Description

Synonym 2-Carboxy-3'-fluorobiphenyl, 2-(3-Fluorophenyl)benzoic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 96-98
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-11-biphenyl-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-11-biphenyl-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart