APO455834509 Apollo Scientific Ltd

4'-Fluoro-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number MFCD00452724
  • Cas Number : 10540-39-3
  • Molecular Weight : 216.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Carboxy-4'-fluorobiphenyl, 3-(4-Fluorophenyl)benzoic acid
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-11-biphenyl-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-11-biphenyl-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart