APO455834511 Apollo Scientific Ltd

3'-Fluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number MFCD04039228
  • Cas Number : 22494-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(3-Fluorophenyl)-2-hydroxybenzoic acid, 5-(3-Fluorophenyl)salicyclic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8683

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8683
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart