APO455834512 Apollo Scientific Ltd

4,5-Difluoro-2-hydroxybenzaldehyde

  • MDL number MFCD08458005
  • Cas Number : 199287-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-72
Synonym 4,5-Difluorosalicylaldehdye, 4,5-Difluoro-2-formylphenol
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9638

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9638
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart