APO455834514 Apollo Scientific Ltd

5-(3-Fluorophenyl)-2-furoic acid

  • MDL number MFCD04039229
  • Cas Number : 54022-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8686

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8686
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart