APO455834520 Apollo Scientific Ltd

4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxaldehyde

  • MDL number MFCD01862519
  • Cas Number : 90035-34-0
  • Molecular Weight : 250.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-72
Synonym 4-[4-(Trifluoromethyl)phenyl]benzaldehyde
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethyl-11-biphenyl-4-carboxaldehyde

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethyl-11-biphenyl-4-carboxaldehyde
pictograms GHS07;GHS08
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart