APO455834521 Apollo Scientific Ltd

4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carboxaldehyde

  • MDL number MFCD01631879
  • Cas Number : 343604-24-0
  • Molecular Weight : 250.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[4-(Trifluoromethyl)phenyl]benzaldehyde, 4-(3-Formylphenyl)benzotrifluoride
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethyl-11-biphenyl-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethyl-11-biphenyl-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart