APO455834528 Apollo Scientific Ltd

3-Amino-4-(methylsulphonyl)benzotrifluoride

  • MDL number MFCD04037961
  • Cas Number : 402-19-7
  • Molecular Weight : 239.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Synonym 2-(Methylsulphonyl)-5-(trifluoromethyl)aniline, 2-Amino-4-(trifluoromethyl)phenyl methyl sulphone
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-methylsulphonylbenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-methylsulphonylbenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart