APO455834535 Apollo Scientific Ltd

3,5-Dichloro-4-fluoronitrobenzene

  • MDL number MFCD00129963
  • Cas Number : 3107-19-5
  • Molecular Weight : 209.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44
Synonym 1,3-Dichloro-2-fluoro-5-nitrobenzene
Purity 99
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dichloro-4-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloro-4-fluoronitrobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart