APO455834540 Apollo Scientific Ltd

3',5'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4-sulphonyl chloride

  • MDL number MFCD01631924
  • Cas Number : 885950-95-8
  • Molecular Weight : 388.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[3,5-Bis(trifluoromethyl)phenyl]benzenesulphonyl chloride, 3',5'-Bis(trifluoromethyl)-4-(chlorosulphonyl)biphenyl
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-bistrifluoromethyl-11-biphenyl-4-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/35-bistrifluoromethyl-11-biphenyl-4-sulphonyl-chloride
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart