APO455834544 Apollo Scientific Ltd

4-Bromo-2-fluorobenzotrifluoride

  • MDL number MFCD01075254
  • Cas Number : 142808-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-fluoro-1-(trifluoromethyl)benzene, 4-Bromo-alpha,alpha,alpha,2-tetrafluorotoluene
Density 1.72
Flash Point 87
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-fluorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart