APO455834545 Apollo Scientific Ltd

3-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline

  • MDL number MFCD03407378
  • Cas Number : 59108-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-2-(trifluoromethyl)quinolin-4-ol, 3-Bromo-4-hydroxy-2-(trifluoromethyl)-1-azanaphthalene
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8921

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8921
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart