APO455834553 Apollo Scientific Ltd

N,N-Dimethyl-2-(trifluoromethoxy)benzylamine

  • MDL number MFCD07368741
  • Cas Number : 886762-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethyl-1-[2-(trifluoromethoxy)phenyl]methylamine
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9274

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9274
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart