APO455834561 Apollo Scientific Ltd

3,6-Difluoro-2-hydroxybenzoic acid

  • MDL number MFCD08275285
  • Cas Number : 749230-37-3
  • Molecular Weight : 174.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-155
Synonym 3,6-Difluorosalicylic acid, 2-Carboxy-3,6-difluorophenol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/36-difluoro-2-hydroxybenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/36-difluoro-2-hydroxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart