APO455834562 Apollo Scientific Ltd

3,5-Difluoro-2-hydroxybenzoic acid

  • MDL number MFCD06203609
  • Cas Number : 84376-20-5
  • Molecular Weight : 174.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 199-203
Iupac Name 3,5-difluoro-2-hydroxybenzoic acid
Inchi Code 1S/C7H4F2O3/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2,10H,(H,11,12)
Inchi Key GZPCNALAXFNOBT-UHFFFAOYSA-N

Description

Synonym 3,5-Difluorosalicylic acid, 2-Carboxy-4,6-difluorophenol
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 199-203
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-2-hydroxybenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-2-hydroxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart