APO455834564 Apollo Scientific Ltd

2,5-Difluoro-4-methoxybenzonitrile

  • MDL number MFCD06246886
  • Cas Number : 1007605-44-8
  • Molecular Weight : 169.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-93
Synonym 4-Cyano-2,5-difluoroanisole, 2,5-Difluoro-p-anisonitrile
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-difluoro-4-methoxybenzonitrile

Safety Information

signalword Danger
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/25-difluoro-4-methoxybenzonitrile
pictograms GHS06
hstatements H302+H332;H311;H319
Availability details for
Quantity
  • Availability
  • Add to Cart