APO455834572 Apollo Scientific Ltd

Hexakis(2,2-difluoroethoxy)phosphazene

  • MDL number MFCD07784333
  • Cas Number : 186817-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-42
Synonym 2,2,4,4,6,6-Hexakis(2,2-difluoroethoxy)-1,3,5-triazatriphosphinine
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33441

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33441
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart