APO455834580 Apollo Scientific Ltd

3,4-Difluoro-5-methoxybenzeneboronic acid

  • MDL number MFCD09907960
  • Cas Number : 925910-42-5
  • Molecular Weight : 187.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-difluoro-5-methoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/34-difluoro-5-methoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart