APO455834582 Apollo Scientific Ltd

4-Bromo-2,6-difluorophenol

  • MDL number MFCD00236213
  • Cas Number : 104197-13-9
  • Molecular Weight : 208.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-26-difluorophenol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-26-difluorophenol
pictograms GHS06;GHS07
hstatements H301+H311+H331;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart