APO455834593 Apollo Scientific Ltd

2,2'-(Benzene-1,4-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)

  • MDL number MFCD00042091
  • Cas Number : 1992-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87
Synonym 1,4-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxyprop-2-yl)benzene, 1,4-Bis[perfluoro(2-hydroxyprop-2-yl)]benzene
Purity 97
Density 1.659
Flash Point 108
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 99/20mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=248

Safety Information

signalword Warning
pcodes P391;P501
MSDS http://www.apolloscientific.co.uk/display_item.php?id=248
pictograms GHS07;GHS09
hstatements H315;H319;H335;H411
Availability details for
Quantity
  • Availability
  • Add to Cart