APO455834595 Apollo Scientific Ltd

5,6,7,8-Tetrahydro-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD08460920
  • Cas Number : 486460-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Trifluoromethyl)[1,2,4]triazolo[4,3-a]piperazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5678-tetrahydro-3-trifluoromethyl124triazolo43-apyrazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5678-tetrahydro-3-trifluoromethyl124triazolo43-apyrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart