APO455834599 Apollo Scientific Ltd

2-Fluoro-5-hydroxypyridine

  • MDL number MFCD01075040
  • Cas Number : 55758-32-2
  • Molecular Weight : 113.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 145-148
Iupac Name 6-fluoro-3-pyridinol
Inchi Code 1S/C5H4FNO/c6-5-2-1-4(8)3-7-5/h1-3,8H
Inchi Key HTRLNWYWOKWCLV-UHFFFAOYSA-N

Description

Synonym 6-Fluoropyridin-3-ol
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 145-148
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart