APO455834600 Apollo Scientific Ltd

4-Chloro-2-fluorobenzyl chloride

  • MDL number MFCD03412453
  • Cas Number : 87417-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-1-(chloromethyl)-2-fluorobenzene, alpha,4-Dichloro-2-fluorotoluene
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9761

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9761
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart