APO455834601 Apollo Scientific Ltd

Gallium(III) trifluoromethanesulphonate

  • MDL number MFCD02093898
  • Cas Number : 74974-60-0
  • Molecular Weight : 516.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Gallium(III) triflate
Density 1.7
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/galliumiii-trifluoromethanesulphonate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/galliumiii-trifluoromethanesulphonate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart