APO455834609 Apollo Scientific Ltd

N-[(2,3-Difluorophenyl)carbamothioyl]benzamide

  • MDL number MFCD08458007
  • Cas Number : 887267-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-143
Synonym 1-Benzoyl-3-(2,3-difluorophenyl)thiourea
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9723

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9723
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart