APO455834622 Apollo Scientific Ltd

4-Bromo-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD03095347
  • Cas Number : 1735-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-5-cyanobenzotrifluoride, 4-Bromo-alpha,alpha,alpha-trifluoro-m-tolunitrile
Purity 97
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylbenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylbenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart