APO455834625 Apollo Scientific Ltd

2-Chloro-4-fluorobromobenzene

  • MDL number MFCD00142587
  • Cas Number : 110407-59-5
  • Molecular Weight : 209.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-chloro-4-fluorobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluorobromobenzene

Safety Information

signalword Danger
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluorobromobenzene
pictograms GHS07;GHS08
hstatements H319;H334;H335
Availability details for
Quantity
  • Availability
  • Add to Cart