APO455834629 Apollo Scientific Ltd

3-Bromo-4-chlorobenzotrifluoride

  • MDL number MFCD00018093
  • Cas Number : 454-78-4
  • Molecular Weight : 259.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -23 to -22
Synonym 3-Bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene, 2-Bromo-1-chloro-4-(trifluoromethyl)benzene
Purity 97
Density 1.72
Flash Point 94
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 188-192
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-chlorobenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-chlorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart