APO455834630 Apollo Scientific Ltd

4-Bromo-3-fluoroanisole

  • MDL number MFCD01310983
  • Cas Number : 458-50-4
  • Molecular Weight : 205.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-fluoro-4-methoxybenzene, 4-Bromo-3-fluorophenyl methyl ether
Purity 98
Density 1.60
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 216
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-fluoroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-fluoroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart