APO455834639 Apollo Scientific Ltd

4-Chloro-3-fluorobromobenzene

  • MDL number MFCD00042519
  • Cas Number : 60811-18-9
  • Molecular Weight : 209.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-1-chloro-2-fluorobenzene
Purity 98
Density 1.71
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 62-64/9mm
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-fluorobromobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-fluorobromobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart