APO455834641 Apollo Scientific Ltd

2-Bromo-4-fluoro-5-methylbenzonitrile

  • MDL number MFCD07782070
  • Cas Number : 916792-07-9
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-5-cyano-2-fluorotoluene, 6-Bromo-4-fluoro-m-tolunitrile
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-5-methylbenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-fluoro-5-methylbenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart