APO455834649 Apollo Scientific Ltd

3-Bromo-2-fluorophenylacetic acid

  • MDL number MFCD09864663
  • Cas Number : 1375964-39-8
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-135
Synonym (3-Bromo-2-fluorophenyl)acetic acid, 2-(3-Bromo-2-fluorophenyl)ethanoic acid
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-fluorophenylacetic-acid

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-fluorophenylacetic-acid
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart