APO455834667 Apollo Scientific Ltd

3-Fluoro-2-nitrobenzoic acid

  • MDL number MFCD07368342
  • Cas Number : 1000339-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-195
Synonym 2-Carboxy-6-fluoronitrobenzene
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40838

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40838
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart