APO455834676 Apollo Scientific Ltd

2-Chloro-3-fluoro-5-methylpyridine

  • MDL number MFCD06658238
  • Cas Number : 34552-15-3
  • Molecular Weight : 145.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Chloro-5-fluoro-3-picoline
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-fluoro-5-methylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-fluoro-5-methylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart