APO455834677 Apollo Scientific Ltd

3-Fluoro-2-nitrobenzonitrile

  • MDL number MFCD09864664
  • Cas Number : 1000339-52-5
  • Molecular Weight : 166.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-nitrobenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-nitrobenzonitrile
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart