APO455834678 Apollo Scientific Ltd

3,4,5,6-Tetrafluorophthalimide

  • MDL number MFCD00091519
  • Cas Number : 652-11-9
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5,6,7-tetrafluoro-1H-isoindole-1,3(2H)-dione
Inchi Code 1S/C8HF4NO2/c9-3-1-2(8(15)13-7(1)14)4(10)6(12)5(3)11/h(H,13,14,15)
Inchi Key GXHIOZHDNIONPT-UHFFFAOYSA-N

Description

Synonym 4,5,6,7-Tetrafluoro-1H-isoindole-1,3(2H)-dione, 4,5,6,7-Tetrafluoroisoindoline-1,3-dione, 1,3-Dioxo-4,5,6,7-tetrafluoroisoindoline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3456-tetrafluorophthalimide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3456-tetrafluorophthalimide
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart