APO455834680 Apollo Scientific Ltd

1-[2-(Trifluoromethoxy)benzyl]piperazine

  • MDL number MFCD07368743
  • Cas Number : 886762-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[(Piperazin-1-yl)methyl]-2-(trifluoromethoxy)benzene
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 78-80/0.02mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8826

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8826
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart