APO455834681 Apollo Scientific Ltd

1-[4-(Trifluoromethoxy)benzyl]piperazine

  • MDL number MFCD07368745
  • Cas Number : 340759-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[(Piperazin-1-yl)methyl]-4-(trifluoromethoxy)benzene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8829

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8829
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart