APO455834687 Apollo Scientific Ltd

4-Bromo-3-fluorobenzotrifluoride

  • MDL number MFCD00042497
  • Cas Number : 40161-54-4
  • Molecular Weight : 243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-fluoro-4-(trifluoromethyl)benzene, 4-Bromo-alpha,alpha,alpha,3-tetrafluorotoluene
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 154-155
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-fluorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart